Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Fisherbrand
Sibutramine hydrochloride monohydrate
CAS: 125494-59-9 | C17H29Cl2NO | 334.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA462200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSibutramine hydrochloride monohydrate
CAS125494-59-9
Molecular FormulaC17H29Cl2NO
Quantity1 g
InChI KeyKFNNPQDSPLWLCX-UHFFFAOYNA-N