Search Fisher Scientific
| Catalog Number | Capacity |
|---|---|
| 010-1072 | 30 mL Oak Ridge Tube |
| 010-1074 | 16 mL Oak Ridge Tube |
| 010-1410 | 15 mL Conical Tube |
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.