Dowex™ 1X8, 200-400 mesh, ion-exchange resin
Dowex™ 1X8, 200-400 mesh, ion-exchange resin
Dowex™ 1X8, 200-400 mesh, ion-exchange resin
Acros Organics
Dowex™ 1X8, 200-400 mesh, ion-exchange resin
CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR203015000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 1X8
Physical FormMoist Beads
CAS12627-85-9
Particle Size15 % max. (on 100 mesh, cumulative) 15 % max. (through 325 mesh)
PackagingPlastic bottle