DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals
Acros Organics
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR450710050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Glyceraldehyde dimer
CAS23147-59-3
Melting Point138.0°C
Infrared SpectrumAuthentic
Assay Percent Range94% min. (GC)